Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitroisophthalic acid monomethyl ester, 99%

$77.11 - $280.15

Chemical Identifiers

Chemical Name or Material 5-Nitroisophthalic acid monomethyl ester
CAS 1955-46-0
Molecular Formula C9H7NO6
Formula Weight 225.16
MDL Number MFCD00009793
Beilstein 1469253
Synonym Methyl hydrogen 5-nitroisophthalate; Monomethyl 5-nitrobenzene-1,3-dicarboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1652518

 
alfa aesar™
A1652518
50g Each for $77.11

AAA1652530

 
alfa aesar™
A1652530
250g Each for $280.15
Description & Specifications

Specifications

Formula Weight 225.16
Chemical Name or Material 5-Nitroisophthalic acid monomethyl ester
TSCA TSCA
Assay 99%
Molecular Formula C9H7NO6
MDL Number MFCD00009793
CAS 1955-46-0
Beilstein 1469253
Synonym Methyl hydrogen 5-nitroisophthalate; Monomethyl 5-nitrobenzene-1,3-dicarboxylate
Melting Point 180° to 185°C
Quantity 50g