Learn More
5-Nitroisophthalic acid, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1169622
Description
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
Solubility
Soluble in water.1.5 g/L at 20°C
Notes
Incompatible with strong oxidizing agents, strong bases.
Specifications
5-Nitroisophthalic acid | |
260°C to 264°C | |
C8H5NO6 | |
100 g | |
5-nitroisophthalic acid, 5-nitrobenzene-1,3-dicarboxylic acid, 5-nitro-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylic acid, 5-nitro, 5-nitroisophthalicacid, 5-nitro-m-phthalic acid, unii-g7rz7lc76u, isophthalic acid, 5-nitro, 1-nitrobenzene-3,5-dicarboxylic acid, g7rz7lc76u | |
NBDAHKQJXVLAID-UHFFFAOYSA-N | |
211.129 | |
211.13 |
618-88-2 | |
Odorless | |
MFCD00007254 | |
1976587 | |
Soluble in water.1.5g/L at 20°C | |
C1=C(C=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O | |
12069 | |
98% |
Chemical Identifiers
618-88-2 | |
211.129 | |
NBDAHKQJXVLAID-UHFFFAOYSA-N | |
12069 |
C8H5NO6 | |
MFCD00007254 | |
5-nitroisophthalic acid, 5-nitrobenzene-1,3-dicarboxylic acid, 5-nitro-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylic acid, 5-nitro, 5-nitroisophthalicacid, 5-nitro-m-phthalic acid, unii-g7rz7lc76u, isophthalic acid, 5-nitro, 1-nitrobenzene-3,5-dicarboxylic acid, g7rz7lc76u | |
C1=C(C=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Warning
EINECSNumber : 210-568-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only