missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nitroisatin, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2496214

 View more versions of this product

Catalog No. AAB2496214



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Nitroisatin
∼252°C (decomposition)
MFCD00005720
180223
UNMYHYODJHKLOC-UHFFFAOYSA-N
5-nitro-1H-indole-2,3-dione
4669250
98%
611-09-6
C8H4N2O4
25 g
5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2
192.13
192.13

Chemical Identifiers

611-09-6
192.13
UNMYHYODJHKLOC-UHFFFAOYSA-N
4669250
C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2
C8H4N2O4
MFCD00005720
5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
5-nitro-1H-indole-2,3-dione
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302-H312-H332-H340

EINECSNumber : 210-252-5

RTECSNumber : NL7970000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only