Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitroindoline, 97%

$113.31 - $414.36

Chemical Identifiers

Chemical Name or Material 5-Nitroindoline
CAS 32692-19-6
Molecular Formula C8H8N2O2
Formula Weight 164.16
MDL Number MFCD00005709
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2541706

  SDS
alfa aesar™
B2541706
5g Each for $113.31

AAB2541714

  SDS
alfa aesar™
B2541714
25g Each for $414.36
Description & Specifications

Specifications

Formula Weight 164.16
Chemical Name or Material 5-Nitroindoline
Assay 97%
Molecular Formula C8H8N2O2
MDL Number MFCD00005709
CAS 32692-19-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 92° to 94°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.