Please login to your online account to display your discounted pricing

5-Nitroindole 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  128700050

 View more versions of this product

Catalog No. AC128700050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder or Needles
Formula Weight 162.15
Quantity 5g
Color Yellow
EINECS Number 228-153-0
RTECS Number NM1168200
Melting Point 140° to 143°C
CAS Max % 100.0
Chemical Name or Material 5-Nitroindole
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H6N2O2
MDL Number MFCD00005673
CAS 6146-52-7
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 5-Nitroindole
Name Note 99%
CAS 6146-52-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00005673
Accessories

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.

SDS & Safety Documents