missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Nitroindole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1060806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Nitroindole | |
139°C to 143°C | |
MFCD00005673 | |
383777 | |
OZFPSOBLQZPIAV-UHFFFAOYSA-N | |
5-nitro-1H-indole | |
22523 | |
99% |
6146-52-7 | |
C8H6N2O2 | |
5 g | |
5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr | |
C1=CC2=C(C=CN2)C=C1[N+](=O)[O-] | |
162.148 | |
162.15 |
Chemical Identifiers
6146-52-7 | |
162.148 | |
OZFPSOBLQZPIAV-UHFFFAOYSA-N | |
22523 | |
C1=CC2=C(C=CN2)C=C1[N+](=O)[O-] |
C8H6N2O2 | |
MFCD00005673 | |
5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr | |
5-nitro-1H-indole |
Safety and Handling
GHS H Statement
H341
Suspected of causing genetic defects.
P201-P202-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c
H302-H319-H340
EINECSNumber : 228-153-0
RTECSNumber : NM1168200
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only