Please login to your online account to display your discounted pricing

5-Nitro-m-xylene ≥99%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  160180500

 View more versions of this product

Catalog No. AC160180500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 151.16
Physical Form Crystals or Chunks
Quantity 50g
Color Yellow to yellow-orange
EINECS Number 202-732-8
TSCA TSCA
Melting Point 68° to 74°C
Beilstein 05, 378
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Packaging Glass bottle
Synonym 1, 3-Dimethyl-5-nitrobenzene
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
CAS 99-12-7
Linear Formula (CH3)2C6H3NO2
Assay Percent Range 97%
Molecular Formula C8H9NO2
Chemical Name or Material 5-Nitro-m-xylene
MDL Number MFCD00007269
Name Note 99+%

Specifications

Chemical Name or Material 5-Nitro-m-xylene
Name Note 99+%
CAS 99-12-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9NO2
Linear Formula (CH3)2C6H3NO2
Formula Weight 151.16
MDL Number MFCD00007269
Synonym 1, 3-Dimethyl-5-nitrobenzene
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

SDS & Safety Documents