Please login to your online account to display your discounted pricing

5-Nitro-m-xylene ≥99%, ACROS Organics™

$211.09 - $211.09

Chemical Identifiers

Chemical Name or Material 5-Nitro-m-xylene
Name Note 99+%
CAS 99-12-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9NO2
Linear Formula (CH3)2C6H3NO2
Formula Weight 151.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160180500

  SDS
thermo fisher scientific
160180500
Glass bottle 50g Each for $211.09
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystals or Chunks
Formula Weight 151.16
Quantity 50g
Color Yellow to yellow-orange
EINECS Number 202-732-8
TSCA TSCA
Melting Point 68° to 74°C
Beilstein 05, 378
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Infrared Spectrum Authentic
Chemical Name or Material 5-Nitro-m-xylene
Name Note 99+%
Assay Percent Range 97%
Molecular Formula C8H9NO2
Linear Formula (CH3)2C6H3NO2
MDL Number MFCD00007269
CAS 99-12-7
Synonym 1, 3-Dimethyl-5-nitrobenzene
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.