Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-m-xylene, 99%

Chemical Identifiers

Chemical Name or Material 5-Nitro-m-xylene
CAS 99-12-7
Molecular Formula C8H9NO2
Formula Weight 151.17
MDL Number MFCD00007269
UN Number UN3447
Beilstein 1936130
Synonym 1,3-Dimethyl-5-nitrobenzene; 3,5-Dimethylnitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0776209

  SDS
alfa aesar™
L0776209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0776218

  SDS
alfa aesar™
L0776218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 151.17
Assay 99%
Chemical Name or Material 5-Nitro-m-xylene
TSCA TSCA
Molecular Formula C8H9NO2
MDL Number MFCD00007269
CAS 99-12-7
UN Number UN3447
Beilstein 1936130
Synonym 1,3-Dimethyl-5-nitrobenzene; 3,5-Dimethylnitrobenzene
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROXYLENES, SOLID
Boiling Point 273°C
Melting Point 69° to 73°C
Quantity 10g
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

Applications
5-Nitro-m-xylene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.