missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-6-methyluracil, 99%

Manufacturer:  Alfa Aesar™ H5591303

 View more versions of this product

Catalog No. AAH5591303



Description

Description

Applications
5-Nitro-6-methyluracil, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in DMF, DMSO, hot methanol and hot water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

5-Nitro-6-methyluracil
16632-21-6
MFCD00047362
Soluble in DMF,DMSO,hot methanol and hot water.
CC1=C(C(=O)NC(=O)N1)[N+](=O)[O-]
171.112
171.11
1g
C5H5N3O4
2,4 1h,3h-pyrimidinedione, 6-methyl-5-nitro
LIVYMRJSNFHYEN-UHFFFAOYSA-N
6-methyl-5-nitro-1H-pyrimidine-2,4-dione
85523
99%

Chemical Identifiers

16632-21-6
171.112
LIVYMRJSNFHYEN-UHFFFAOYSA-N
85523
CC1=C(C(=O)NC(=O)N1)[N+](=O)[O-]
C5H5N3O4
MFCD00047362
2,4 1h,3h-pyrimidinedione, 6-methyl-5-nitro
6-methyl-5-nitro-1H-pyrimidine-2,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

EINECSNumber : 240-688-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents