missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-2-(n-propylamino)pyridine, 96%

$55.35 - $294.70

Chemical Identifiers

CAS 25948-11-2
Molecular Formula C8H11N3O2
Formula Weight 181.19
MDL Number MFCD00084819
Chemical Name or Material 5-Nitro-2-(n-propylamino)pyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2741603 SDS Alfa Aesar™
H2741603
1g Each for $55.35
AAH2741609 SDS Alfa Aesar™
H2741609
10g Each for $294.70
Specifications Description & Specifications

Chemical Identifiers

25948-11-2
181.19
5-Nitro-2-(n-propylamino)pyridine
C8H11N3O2
MFCD00084819

Specifications

94° to 96°C
25948-11-2
181.19
5-Nitro-2-(n-propylamino)pyridine
1g
C8H11N3O2
MFCD00084819
96%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger