Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-2-furoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 5-Nitro-2-furoyl chloride
CAS 25084-14-4
Molecular Formula C5H2ClNO4
Formula Weight 175.53
MDL Number MFCD00039564
UN Number UN3261
Beilstein 139693
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0146803

  SDS
alfa aesar™
L0146803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0146806

  SDS
alfa aesar™
L0146806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0146814

  SDS
alfa aesar™
L0146814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.53
CAS 25084-14-4
Chemical Name or Material 5-Nitro-2-furoyl chloride
Assay 97%
Molecular Formula C5H2ClNO4
Beilstein 139693
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 137° to 140°C (15mmHg)
Quantity 1g
MDL Number MFCD00039564
UN Number UN3261
Melting Point 38° to 42°C
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.