missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-2-furoic acid, 98+%

$41.32 - $544.36

Chemical Identifiers

CAS 645-12-5
Molecular Formula C5H3NO5
Molecular Weight (g/mol) 157.081
MDL Number MFCD00003240
InChI Key IODMEDPPCXSFLD-UHFFFAOYSA-N
Synonym 5-nitro-2-furoic acid, nitrofurate, 5-nitrofuroic acid, 2-furoic acid, 5-nitro, 5-nitro-2-furancarboxylic acid, 5-nitropyromucate, 5-nitrofurancarboxylic acid, 2-furancarboxylic acid, 5-nitro, ccris 1197, 5-nitropyromucic acid
PubChem CID 12577
IUPAC Name 5-nitrofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)[N+](=O)[O-])C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1059706 View Documents Alfa Aesar™
A1059706
5g Each for $41.32
 
AAA1059714 View Documents Alfa Aesar™
A1059714
25g Each for $163.46
 
AAA1059722 View Documents Alfa Aesar™
A1059722
100g Each for $544.36
 
Description

Description

Applications
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

645-12-5
157.081
IODMEDPPCXSFLD-UHFFFAOYSA-N
12577
C1=C(OC(=C1)[N+](=O)[O-])C(=O)O
C5H3NO5
MFCD00003240
5-nitro-2-furoic acid, nitrofurate, 5-nitrofuroic acid, 2-furoic acid, 5-nitro, 5-nitro-2-furancarboxylic acid, 5-nitropyromucate, 5-nitrofurancarboxylic acid, 2-furancarboxylic acid, 5-nitro, ccris 1197, 5-nitropyromucic acid
5-nitrofuran-2-carboxylic acid

Specifications

186°C to 188°C
645-12-5
MFCD00003240
5-nitro-2-furoic acid, nitrofurate, 5-nitrofuroic acid, 2-furoic acid, 5-nitro, 5-nitro-2-furancarboxylic acid, 5-nitropyromucate, 5-nitrofurancarboxylic acid, 2-furancarboxylic acid, 5-nitro, ccris 1197, 5-nitropyromucic acid
C1=C(OC(=C1)[N+](=O)[O-])C(=O)O
157.081
157.08
5-Nitro-2-furoic acid
5g
C5H3NO5
139373
IODMEDPPCXSFLD-UHFFFAOYSA-N
5-nitrofuran-2-carboxylic acid
12577
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LV2012000