missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-2-furaldoxime, 97%

$39.47 - $105.95

Chemical Identifiers

CAS 555-15-7
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.097
MDL Number MFCD00198004
InChI Key PTBKFATYSVLSSD-ZZXKWVIFSA-N
Synonym nifuroxime, 5-nitro-2-furaldehyde oxime, usaf ea-5, 2-furaldehyde, 5-nitro-, oxime, 2-furaldoxime, 5-nitro, 5-nitro-2-furaldoxime, carbazylquinone, carbilquinone, 5-nitrofuran-2-carbaldehyde oxime, 5-nitro-2-furancarboxaldehyde oxime
PubChem CID 6478035
IUPAC Name (NE)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=NO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0137409 View Documents Alfa Aesar™
L0137409
10g Each for $39.47
 
AAL0137418 View Documents Alfa Aesar™
L0137418
50g Each for $105.95
 
Specifications

Chemical Identifiers

555-15-7
156.097
PTBKFATYSVLSSD-ZZXKWVIFSA-N
6478035
C1=C(OC(=C1)[N+](=O)[O-])C=NO
C5H4N2O4
MFCD00198004
nifuroxime, 5-nitro-2-furaldehyde oxime, usaf ea-5, 2-furaldehyde, 5-nitro-, oxime, 2-furaldoxime, 5-nitro, 5-nitro-2-furaldoxime, carbazylquinone, carbilquinone, 5-nitrofuran-2-carbaldehyde oxime, 5-nitro-2-furancarboxaldehyde oxime
(NE)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine

Specifications

10g
C5H4N2O4
83037
nifuroxime, 5-nitro-2-furaldehyde oxime, usaf ea-5, 2-furaldehyde, 5-nitro-, oxime, 2-furaldoxime, 5-nitro, 5-nitro-2-furaldoxime, carbazylquinone, carbilquinone, 5-nitrofuran-2-carbaldehyde oxime, 5-nitro-2-furancarboxaldehyde oxime
C1=C(OC(=C1)[N+](=O)[O-])C=NO
156.097
156.1
5-Nitro-2-furaldoxime
555-15-7
MFCD00198004
14,6536
PTBKFATYSVLSSD-ZZXKWVIFSA-N
(NE)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
6478035
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : LT7525000