missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Nitro-2-furaldehyde diacetate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1816936
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Nitro-2-furaldehyde diacetate | |
87°C to 92°C | |
MFCD00003244 | |
255089 | |
5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate | |
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C | |
243.171 | |
243.17 |
92-55-7 | |
C9H9NO7 | |
500 g | |
Light sensitive | |
HSXKWKJCZNRMJO-UHFFFAOYSA-N | |
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate | |
7097 | |
≥98% |
Chemical Identifiers
92-55-7 | |
243.171 | |
HSXKWKJCZNRMJO-UHFFFAOYSA-N | |
7097 | |
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C |
C9H9NO7 | |
MFCD00003244 | |
5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate | |
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate |
Safety and Handling
GHS H Statement
H341
Suspected of causing genetic defects.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 202-166-1
RTECSNumber : LU1940000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only