missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nitro-2-furaldehyde, 97%, Thermo Scientific™

$86.17 - $86.17

Chemical Identifiers

CAS 698-63-5
Molecular Formula C5H3NO4
Molecular Weight (g/mol) 141.08
MDL Number MFCD00003230
InChI Key SXINBFXPADXIEY-UHFFFAOYSA-N
Synonym 2-furaldehyde, 5-nitro, 2-furancarboxaldehyde, 5-nitro, 5-nitro-2-furaldehyde, 5-nitro-2-furancarboxaldehyde, 5-nitro-2-furfural, 5-nitrofuraldehyde, 5-nitrofurfural, 5-nitrofurfuraldehyde, furfural, 5-nitro, nitrofurfural
PubChem CID 12774
ChEBI CHEBI:34459
IUPAC Name 5-nitrofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC173860100
SDS View Documents Promotion Details
Thermo Fisher Scientific
173860100
10g Glass bottle
Each for $86.17
 
AC173860500
SDS View Documents Promotion Details
Thermo Fisher Scientific
173860500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

698-63-5
141.08
SXINBFXPADXIEY-UHFFFAOYSA-N
12774
5-nitrofuran-2-carbaldehyde
C5H3NO4
MFCD00003230
2-furaldehyde, 5-nitro, 2-furancarboxaldehyde, 5-nitro, 5-nitro-2-furaldehyde, 5-nitro-2-furancarboxaldehyde, 5-nitro-2-furfural, 5-nitrofuraldehyde, 5-nitrofurfural, 5-nitrofurfuraldehyde, furfural, 5-nitro, nitrofurfural
CHEBI:34459
C1=C(OC(=C1)[N+](=O)[O-])C=O

Specifications

698-63-5
100.0
Yellow to Brown
33°C
C5H3NO4
MFCD00003230
17,IV,4459
10g
Solubility in water: slightly soluble
SXINBFXPADXIEY-UHFFFAOYSA-N
5-nitrofuran-2-carbaldehyde
12774
141.08
97%
96.0
1.3400g/mL
34.0°C to 35.0°C
96% min. (GC)
Authentic
Glass bottle
1.5905
2-furaldehyde, 5-nitro, 2-furancarboxaldehyde, 5-nitro, 5-nitro-2-furaldehyde, 5-nitro-2-furancarboxaldehyde, 5-nitro-2-furfural, 5-nitrofuraldehyde, 5-nitrofurfural, 5-nitrofurfuraldehyde, furfural, 5-nitro, nitrofurfural
1.34
C1=C(OC(=C1)[N+](=O)[O-])C=O
141.08
CHEBI:34459
Crystalline Low Melting Solid
5-Nitro-2-furaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.

GHS Signal Word: Warning

EINECSNumber : 211-816-3

SDS
Product Certifications

RUO – Research Use Only