missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nitro-2-furaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC173860500

 View more versions of this product

Catalog No. AC173860500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Nitro-2-furaldehyde
33°C
Glass bottle
1.34
34.0°C to 35.0°C
698-63-5
100.0
C5H3NO4
17,IV,4459
Solubility in water: slightly soluble
C1=C(OC(=C1)[N+](=O)[O-])C=O
141.08
CHEBI:34459
Crystalline Low Melting Solid
1.3400g/mL
Authentic
1.5905
Yellow to Brown
50g
96.0
96% min. (GC)
MFCD00003230
2-furaldehyde, 5-nitro
SXINBFXPADXIEY-UHFFFAOYSA-N
5-nitrofuran-2-carbaldehyde
12774
141.08
97%

Chemical Identifiers

698-63-5
141.08
SXINBFXPADXIEY-UHFFFAOYSA-N
12774
5-nitrofuran-2-carbaldehyde
C5H3NO4
MFCD00003230
2-furaldehyde, 5-nitro
CHEBI:34459
C1=C(OC(=C1)[N+](=O)[O-])C=O
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.

GHS Signal Word: Warning

EINECSNumber : 211-816-3

SDS Documents