Please login to your online account to display your discounted pricing

5-Nitro-2-furaldehyde, 97%, ACROS Organics™


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  AC17386DA

 View more versions of this product

Catalog No. AC17386DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Low Melting Solid
Assay ≥96.0%
Formula Weight 141.08
Quantity 1g
Color Yellow to brown
Density 1.3400g/mL
Melting Point 33° to 37°C
CAS Min % 96.0
CAS Max % 100.0
Synonym 5-Nitrofurfural
Chemical Name or Material 5-Nitro-2-furaldehyde
Assay Percent Range 97%
MDL Number MFCD00003230
CAS 698-63-5
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Flammable solid.
Packaging Amber glass bottle
Molecular Formula C5H3NO4
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Store in a well-ventilated place. Keep container tightly closed.

Specifications

Chemical Name or Material 5-Nitro-2-furaldehyde
CAS 698-63-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H3NO4
Formula Weight 141.08
MDL Number MFCD00003230
Synonym 5-Nitrofurfural
Accessories

Warning

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Store in a well-ventilated place. Keep container tightly closed.

GHS P Statement
Flammable solid.