missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Nitro-2-furaldehyde, 97%, ACROS Organics™

Manufacturer:  Acros Organics 173860100

 View more versions of this product

Catalog No. AC173860100



Specifications

Specifications

5-Nitro-2-furaldehyde
33°C
Glass bottle
Yellow to Brown
10g
96.0
97%
MFCD00003230
SXINBFXPADXIEY-UHFFFAOYSA-N
5-nitrofuran-2-carbaldehyde
12774
141.08
97%
1.3400g/mL
Authentic
1.5905
34.0°C to 35.0°C
698-63-5
100.0
C5H3NO4
5-nitro-2-furaldehyde, 5-nitrofurfural, nitrofurfural, 2-furancarboxaldehyde, 5-nitro, 5-nitrofurfuraldehyde, 5-nitro-2-furfural, 2-furaldehyde, 5-nitro, 5-nitrofuraldehyde, 5-nitro-2-furancarboxaldehyde, furfural, 5-nitro
C1=C(OC(=C1)[N+](=O)[O-])C=O
141.082
CHEBI:34459
Crystalline Low Melting Solid

Chemical Identifiers

698-63-5
141.082
SXINBFXPADXIEY-UHFFFAOYSA-N
12774
5-nitrofuran-2-carbaldehyde
C5H3NO4
MFCD00003230
5-nitro-2-furaldehyde, 5-nitrofurfural, nitrofurfural, 2-furancarboxaldehyde, 5-nitro, 5-nitrofurfuraldehyde, 5-nitro-2-furfural, 2-furaldehyde, 5-nitro, 5-nitrofuraldehyde, 5-nitro-2-furancarboxaldehyde, furfural, 5-nitro
CHEBI:34459
C1=C(OC(=C1)[N+](=O)[O-])C=O
Safety and Handling

Safety and Handling

GHS P Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Store in a well-ventilated place. Keep container tightly closed.

Warning

SDS Documents