Learn More
5-Nitro-2-benzimidazolinone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1639114
Description
5-Nitro-2-benzimidazolinone is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Nitro-2-benzimidazolinone is used as intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
5-Nitro-2-benzimidazolinone | |
>300°C | |
MFCD00220274 | |
UN2811 | |
5-nitro-2-benzimidazolinone, 5-nitro-1h-benzo d imidazol-2 3h-one, 5-nitrobenzimidazol-2-one, 5-nitro-1,3-dihydro-2h-benzimidazol-2-one, 2-hydroxy-5-nitrobenzimidazole, 5-nitro-2 3h-benzimidazolone, 2h-benzimidazol-2-one, 1,3-dihydro-5-nitro, 2-benzimidazolinone, 5-nitro, 5-nitro-1h-benzimidazol-2-ol, 1,3-dihydro-5-nitro-2h-benzimidazol-2-one | |
DLJZIPVEVJOKHB-UHFFFAOYSA-N | |
5-nitro-2,3-dihydro-1H-1,3-benzodiazol-2-one | |
3725611 | |
99% |
93-84-5 | |
C7H5N3O3 | |
25 g | |
171386 | |
Insoluble in water. | |
[O-][N+](=O)C1=CC=C2NC(=O)NC2=C1 | |
179.14 | |
179.14 |
Chemical Identifiers
93-84-5 | |
179.14 | |
DLJZIPVEVJOKHB-UHFFFAOYSA-N | |
3725611 |
C7H5N3O3 | |
MFCD00220274 | |
5-nitro-2-benzimidazolinone, 5-nitro-1h-benzo d imidazol-2 3h-one, 5-nitrobenzimidazol-2-one, 5-nitro-1,3-dihydro-2h-benzimidazol-2-one, 2-hydroxy-5-nitrobenzimidazole, 5-nitro-2 3h-benzimidazolone, 2h-benzimidazol-2-one, 1,3-dihydro-5-nitro, 2-benzimidazolinone, 5-nitro, 5-nitro-1h-benzimidazol-2-ol, 1,3-dihydro-5-nitro-2h-benzimidazol-2-one | |
[O-][N+](=O)C1=CC=C2NC(=O)NC2=C1 |
Safety and Handling
GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
H302+H312+H332-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 202-282-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only