Please login to your online account to display your discounted pricing

5-Nitro-1H-indole-3-carbaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 5-Nitro-1H-indole-3-carbaldehyde
CAS 6625-96-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD01313781
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RH01174EA

  SDS
fisher scientific company
RH01174EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.16
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
MDL Number MFCD01313781
Chemical Name or Material 5-Nitro-1H-indole-3-carbaldehyde
Assay Percent Range 97%
Molecular Formula C9H6N2O3
CAS 6625-96-3