Please login to your online account to display your discounted pricing

5-Nitro-1H-indole-3-carbaldehyde, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RH01174EA

 View more versions of this product

Catalog No. RH01174EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.16
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
MDL Number MFCD01313781
Chemical Name or Material 5-Nitro-1H-indole-3-carbaldehyde
Assay Percent Range 97%
Molecular Formula C9H6N2O3
CAS 6625-96-3

Specifications

Chemical Name or Material 5-Nitro-1H-indole-3-carbaldehyde
CAS 6625-96-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6N2O3
Formula Weight 190.16
MDL Number MFCD01313781