missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-1,10-phenanthroline, 98%

Manufacturer:  Alfa Aesar™ A1142806

 View more versions of this product

Catalog No. AAA1142806



Description

Description

Applications
5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

5-Nitro-1,10-phenanthroline
5g
C12H7N3O2
196245
PDDBTWXLNJNICS-UHFFFAOYSA-N
5-nitro-1,10-phenanthroline
72790
98%
202°C to 204°C
4199-88-6
MFCD00004981
1,10-phenanthroline, 5-nitro, 5-nitro-1, 10-diazaphenanthrene, nitroferroin, 5-nitropyridino 3,2-h quinoline, acmc-209jmn, phhn01, 5nitro-1, 10-phenanthroline, 5-nitro-1,10-diazaphenanthrene, pddbtwxlnjnics-uhfffaoysa, 5-nitro-1,10-phenanthroline, crystalline
C1=CC2=CC(=C3C=CC=NC3=C2N=C1)[N+](=O)[O-]
225.207
225.21

Chemical Identifiers

4199-88-6
225.207
PDDBTWXLNJNICS-UHFFFAOYSA-N
72790
C1=CC2=CC(=C3C=CC=NC3=C2N=C1)[N+](=O)[O-]
C12H7N3O2
MFCD00004981
1,10-phenanthroline, 5-nitro, 5-nitro-1, 10-diazaphenanthrene, nitroferroin, 5-nitropyridino 3,2-h quinoline, acmc-209jmn, phhn01, 5nitro-1, 10-phenanthroline, 5-nitro-1,10-diazaphenanthrene, pddbtwxlnjnics-uhfffaoysa, 5-nitro-1,10-phenanthroline, crystalline
5-nitro-1,10-phenanthroline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents