Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Nitro-1,10-phenanthroline-2,9-dicarboxylic acid hydrate, 98%

$134.41 - $527.36

Chemical Identifiers

Chemical Name or Material 5-Nitro-1,10-phenanthroline-2,9-dicarboxylic acid hydrate
CAS 164394-23-4
Molecular Formula C14H7N3O6·xH2O
Formula Weight 313.23 (Anhydrous)
MDL Number MFCD00209668
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA18550MC

  SDS
alfa aesar™
A18550MC
100mg Each for $134.41

AAA18550ME

  SDS
alfa aesar™
A18550ME
500mg Each for $527.36
Description & Specifications

Specifications

Formula Weight 313.23 (Anhydrous)
Health Hazard 1 Warning
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈215°C (dec.)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 5-Nitro-1,10-phenanthroline-2,9-dicarboxylic acid hydrate
CAS 164394-23-4
Molecular Formula C14H7N3O6·xH2O
Quantity 100mg
MDL Number MFCD00209668
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.