Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-n-Hexylthio-1,3,4-thiadiazole-2-thiol, 97%

$415.61 - $415.61

Chemical Identifiers

Chemical Name or Material 5-n-Hexylthio-1,3,4-thiadiazole-2-thiol
CAS 4858-28-0
Molecular Formula C8H14N2S3
Formula Weight 234.41
MDL Number MFCD00186165
Synonym 2-n-Hexylthio-5-mercapto-1,3,4-thiadiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2181506

 
alfa aesar™
B2181506
5g Each for $415.61
Description & Specifications

Specifications

Formula Weight 234.41
Chemical Name or Material 5-n-Hexylthio-1,3,4-thiadiazole-2-thiol
Assay 97%
Molecular Formula C8H14N2S3
MDL Number MFCD00186165
CAS 4858-28-0
Synonym 2-n-Hexylthio-5-mercapto-1,3,4-thiadiazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 73° to 76°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.