missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-n-Butylcarbamoyl-2-fluorobenzeneboronic acid, 98%, Thermo Scientific™
$167.49 - $167.49
Chemical Identifiers
CAS | 874289-50-6 |
---|---|
Molecular Formula | C11H15BFNO3 |
Molecular Weight (g/mol) | 239.053 |
MDL Number | MFCD08235104 |
InChI Key | YFCRSIAYLGXFOB-UHFFFAOYSA-N |
Synonym | 5-butylcarbamoyl-2-fluorophenyl boronic acid, n-butyl 3-borono-4-fluorobenzamide, 5-butylcarbamoyl-2-fluorobenzeneboronic acid, 5-butylcarbamoyl-2-fluorophenylboronic acid, acmc-209qmr, 5-butylcarbamoyl-2-fluorophenyl boronicacid, 5-butylcarbamoyl-2-fluoranyl-phenyl boronic acid, 5-butylamino oxo methyl-2-fluorophenyl boronic acid, boronic acid,b-5-butylamino carbonyl-2-fluorophenyl |
PubChem CID | 44717597 |
IUPAC Name | [5-(butylcarbamoyl)-2-fluorophenyl]boronic acid |
SMILES | B(C1=C(C=CC(=C1)C(=O)NCCCC)F)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH53228MD
|
Thermo Scientific Chemicals
H53228MD |
250 mg |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
874289-50-6 | |
239.053 | |
YFCRSIAYLGXFOB-UHFFFAOYSA-N | |
44717597 | |
B(C1=C(C=CC(=C1)C(=O)NCCCC)F)(O)O |
C11H15BFNO3 | |
MFCD08235104 | |
5-butylcarbamoyl-2-fluorophenyl boronic acid, n-butyl 3-borono-4-fluorobenzamide, 5-butylcarbamoyl-2-fluorobenzeneboronic acid, 5-butylcarbamoyl-2-fluorophenylboronic acid, acmc-209qmr, 5-butylcarbamoyl-2-fluorophenyl boronicacid, 5-butylcarbamoyl-2-fluoranyl-phenyl boronic acid, 5-butylamino oxo methyl-2-fluorophenyl boronic acid, boronic acid,b-5-butylamino carbonyl-2-fluorophenyl | |
[5-(butylcarbamoyl)-2-fluorophenyl]boronic acid |
Specifications
874289-50-6 | |
C11H15BFNO3 | |
250 mg | |
YFCRSIAYLGXFOB-UHFFFAOYSA-N | |
[5-(butylcarbamoyl)-2-fluorophenyl]boronic acid | |
44717597 | |
98% |
174°C to 176°C | |
MFCD08235104 | |
5-butylcarbamoyl-2-fluorophenyl boronic acid, n-butyl 3-borono-4-fluorobenzamide, 5-butylcarbamoyl-2-fluorobenzeneboronic acid, 5-butylcarbamoyl-2-fluorophenylboronic acid, acmc-209qmr, 5-butylcarbamoyl-2-fluorophenyl boronicacid, 5-butylcarbamoyl-2-fluoranyl-phenyl boronic acid, 5-butylamino oxo methyl-2-fluorophenyl boronic acid, boronic acid,b-5-butylamino carbonyl-2-fluorophenyl | |
B(C1=C(C=CC(=C1)C(=O)NCCCC)F)(O)O | |
239.053 | |
239.05 | |
5-n-Butylcarbamoyl-2-fluorobenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures