missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylthiophene-2-carboxaldehyde 97.0+%, TCI America™

Manufacturer:  Tci America M091125G

 View more versions of this product

Catalog No. M091125G



Specifications

Specifications

5-Methylthiophene-2-carboxaldehyde
Brown-Yellow
13679-70-4
MFCD00005434
VAUMDUIUEPIGHM-UHFFFAOYSA-N
5-methylthiophene-2-carbaldehyde
61663
Liquid at 20°C
205°C
25g
C6H6OS
2-formyl-5-methylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methyl-2-formylthiophene, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-aldehyde, 5-methylthiophene-2-carboxaldehyde
CC1=CC=C(S1)C=O
126.173
126.17
≥97.0% (GC)

Chemical Identifiers

13679-70-4
126.173
VAUMDUIUEPIGHM-UHFFFAOYSA-N
61663
CC1=CC=C(S1)C=O
C6H6OS
MFCD00005434
2-formyl-5-methylthiophene, 2-thiophenecarboxaldehyde, 5-methyl, 5-methyl-2-formylthiophene, 5-methyl-2-thenaldehyde, 5-methyl-2-thiophencarboxaldehyde, 5-methyl-2-thiophene carboxaldehyde, 5-methyl-2-thiophenecarbaldehyde, 5-methyl-2-thiophenecarboxaldehyde, 5-methylthiophene-2-aldehyde, 5-methylthiophene-2-carboxaldehyde
5-methylthiophene-2-carbaldehyde
Safety and Handling

Safety and Handling

TSCA : Yes

Documents