Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylthiophene-2-carboxaldehyde, 98+%

$68.42 - $254.65

Chemical Identifiers

Chemical Name or Material 5-Methylthiophene-2-carboxaldehyde
CAS 13679-70-4
Molecular Formula C6H6OS
Formula Weight 126.18
MDL Number MFCD00005434
Beilstein 106896
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1111909

  SDS
alfa aesar™
A1111909
10g Each for $68.42

AAA1111918

  SDS
alfa aesar™
A1111918
50g Each for $254.65
Description & Specifications

Specifications

Formula Weight 126.18
Chemical Name or Material 5-Methylthiophene-2-carboxaldehyde
TSCA TSCA
Assay ≥98%
Molecular Formula C6H6OS
MDL Number MFCD00005434
CAS 13679-70-4
Beilstein 106896
Health Hazard 1 Warning
Boiling Point 92° to 93°C (12mmHg)
Quantity 10g
Flash Point 87°C (188°F)
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
5-Methylthiophene-2-carboxaldehyde is used as a food additive. It is an analytical reagent. The reagent has been synthesized and characterized using IR, 1H NMR and Mass spectral data.

Solubility
Soluble in ether and ethanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.