Please login to your online account to display your discounted pricing

5-Methylthiophene-2-carbonitrile, 95%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC65016DE

 View more versions of this product

Catalog No. CC65016DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 123.18
Quantity 5g
Assay 95%
CAS Max % 100.0
Molecular Formula C6H5NS
CAS 72835-25-7
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Health Hazard 2 GHS P Statement Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. Flammable liquid and vapor.
CAS Min % 94.5
Chemical Name or Material 5-Methylthiophene-2-carbonitrile
Packaging Amber Glass Bottle
MDL Number MFCD01434334
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 5-Methylthiophene-2-carbonitrile
CAS 72835-25-7
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C6H5NS
Formula Weight 123.18
MDL Number MFCD01434334
Accessories

GHS Signal Word: Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS P Statement Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. Flammable liquid and vapor.

SDS & Safety Documents