missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylthiophene-2-carbonitrile, 95%, Maybridge

Chemical Identifiers

CAS 72835-25-7
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C6H5NS
Formula Weight 123.18
MDL Number MFCD01434334
Chemical Name or Material 5-Methylthiophene-2-carbonitrile
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC65016DE SDS Fisher Scientific Uk Ltd
CC65016DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC65016DA SDS Fisher Scientific Uk Ltd
CC65016DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

72835-25-7
100.0
C6H5NS
MFCD01434334
95%
94.5
95%
123.18
5-Methylthiophene-2-carbonitrile

Specifications

5g
94.5
Amber Glass Bottle
C6H5NS
MFCD01434334
95%
72835-25-7
100.0
95%
123.18
5-Methylthiophene-2-carbonitrile

Safety and Handling

GHS P Statement Harmful if inhaled. Harmful in contact with skin. Harmful if swallowed. Flammable liquid and vapor.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS Signal Word: Warning