Please login to your online account to display your discounted pricing

5-Methylthiophene-2-boronic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 5-Methylthiophene-2-boronic acid
CAS 162607-20-7
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C5H7BO2S
Formula Weight 141.99g/mol
MDL Number MFCD01318166
Synonym 5-Methyl-2-thienylboric acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC359610010

  SDS
fisher scientific acros
359610010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White to light yellow
Quantity 1g
Chemical Name or Material 5-Methylthiophene-2-boronic acid
CAS 162607-20-7
CAS Min % 97.5
CAS Max % 100.0
Physical Form White to light yellow powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C5H7BO2S
Formula Weight 141.99g/mol
MDL Number MFCD01318166
Synonym 5-Methyl-2-thienylboric acid
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.