missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-(Methylthio)thiophene-2-carboxaldehyde, 97%

$98.55 - $393.24

Chemical Identifiers

CAS 24445-35-0
Molecular Formula C6H6OS2
Molecular Weight (g/mol) 158.233
MDL Number MFCD00173744
InChI Key RKXYTFTZODXDEF-UHFFFAOYSA-N
Synonym 5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
PubChem CID 2736997
IUPAC Name 5-methylsulfanylthiophene-2-carbaldehyde
SMILES CSC1=CC=C(S1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6180303 View Documents Alfa Aesar™
H6180303
1g Each for $98.55
 
AAH6180306 View Documents Alfa Aesar™
H6180306
5g Each for $393.24
 
Specifications

Chemical Identifiers

24445-35-0
158.233
RKXYTFTZODXDEF-UHFFFAOYSA-N
2736997
CSC1=CC=C(S1)C=O
C6H6OS2
MFCD00173744
5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
5-methylsulfanylthiophene-2-carbaldehyde

Specifications

1g
C6H6OS2
5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
CSC1=CC=C(S1)C=O
158.233
158.24
5-(Methylthio)thiophene-2-carboxaldehyde
24445-35-0
MFCD00173744
RKXYTFTZODXDEF-UHFFFAOYSA-N
5-methylsulfanylthiophene-2-carbaldehyde
2736997
97%