missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Methylthio)-1H-tetrazole, TCI America™

$100.83 - $308.63

Chemical Identifiers

CAS 29515-99-9
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.142
MDL Number MFCD00068726
InChI Key ZBXNFTFKKOSPLD-UHFFFAOYSA-N
PubChem CID 97664
IUPAC Name 5-methylsulfanyl-2H-tetrazole
SMILES CSC1=NNN=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18895G View Documents TCI America
M18895G
5g Each for $100.83
 
M188925G View Documents TCI America
M188925G
25g Each for $308.63
 
Specifications

Chemical Identifiers

29515-99-9
116.142
ZBXNFTFKKOSPLD-UHFFFAOYSA-N
5-methylsulfanyl-2H-tetrazole
C2H4N4S
MFCD00068726
97664
CSC1=NNN=N1

Specifications

White-Yellow
5g
C2H4N4S
1325
CSC1=NNN=N1
116.142
116.14
≥98.0% (HPLC)
144°C
29515-99-9
MFCD00068726
ZBXNFTFKKOSPLD-UHFFFAOYSA-N
5-methylsulfanyl-2H-tetrazole
97664
Crystal-Powder at 20°C
5-(Methylthio)-1H-tetrazole
Safety and Handling

Safety and Handling

TSCA : No