Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylthio-1,3,4-thiadiazole-2-thiol, 97%

$41.58 - $41.58

Chemical Identifiers

Chemical Name or Material 5-Methylthio-1,3,4-thiadiazole-2-thiol
CAS 6264-40-0
Molecular Formula C3H4N2S3
Formula Weight 164.27
MDL Number MFCD00051648
Beilstein 4009
Synonym 2-Mercapto-5-methylmercapto-1,3,4-thiadiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0800203

  SDS
alfa aesar™
L0800203
1g Each for $41.58
Description & Specifications

Specifications

Formula Weight 164.27
Chemical Name or Material 5-Methylthio-1,3,4-thiadiazole-2-thiol
CAS 6264-40-0
Assay 97%
MDL Number MFCD00051648
TSCA TSCA
Molecular Formula C3H4N2S3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 134° to 139°C
Health Hazard 1 Warning
Synonym 2-Mercapto-5-methylmercapto-1,3,4-thiadiazole
Beilstein 4009
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.