Please login to your online account to display your discounted pricing

(5-Methylthien-2-yl)methylamine hydrochloride, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC65063DE

 View more versions of this product

Catalog No. CC65063DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 163.67
Quantity 5g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material (5-methylthien-2-yl)methylamine hydrochloride
Packaging Amber Glass Bottle
Molecular Formula C6H9NS·ClH
CAS 171661-55-5
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material (5-methylthien-2-yl)methylamine hydrochloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H9NS·ClH
Formula Weight 163.67
Accessories

GHS Signal Word: Warning