missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylthiazole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438940010

 View more versions of this product

Catalog No. AC438940010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

5-Methylthiazole
Authentic
141°C to 142°C (993.0 hPa)
-40°C
3581-89-3
C4H5NS
RLYUNPNLXMSXAX-UHFFFAOYSA-N
5-methyl-1,3-thiazole
137980
Liquid
42°C
Glass Bottle
Colorless to Yellow
1g
96% min. (GC)
#, 5-methyl thiazole, 5-methyl-thiazole, 5-methylthiazole, acmc-1cjhk, ksc222m5d, thiazole, 5-methyl, zlchem 1239
CC1=CN=CS1
99.16
99.16
97%

Chemical Identifiers

3581-89-3
99.16
#, 5-methyl thiazole, 5-methyl-thiazole, 5-methylthiazole, acmc-1cjhk, ksc222m5d, thiazole, 5-methyl, zlchem 1239
5-methyl-1,3-thiazole
C4H5NS
RLYUNPNLXMSXAX-UHFFFAOYSA-N
137980
CC1=CN=CS1
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a P

GHS Signal Word: Warning

SDS Documents