missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylpyrimidine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450730010

 View more versions of this product

Catalog No. AC450730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-Methylpyrimidine
Glass bottle
33°C to 37°C
2036-41-1
C5H6N2
Solubility in water: soluble
CC1=CN=CN=C1
94.11
94.11
52°C
152°C to 154°C
1g
97.5% min. (GC)
5-methyl-1,3-diazine, 5-methyl-pyrimidin, 5-methyl-pyrimidine, 5-methylpyrimidine, 5-methylpyrimidine, 97+%, acmc-1cnd2, pubchem7207, pyrimidine, 5-methyl, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, rarechem am la 0031
TWGNOYAGHYUFFR-UHFFFAOYSA-N
5-methylpyrimidine
74859
98%

Chemical Identifiers

2036-41-1
94.11
5-methyl-1,3-diazine, 5-methyl-pyrimidin, 5-methyl-pyrimidine, 5-methylpyrimidine, 5-methylpyrimidine, 97+%, acmc-1cnd2, pubchem7207, pyrimidine, 5-methyl, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, rarechem am la 0031
5-methylpyrimidine
C5H6N2
TWGNOYAGHYUFFR-UHFFFAOYSA-N
74859
CC1=CN=CN=C1
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr

GHS Signal Word: Warning

EINECSNumber : 218-006-9

SDS
Documents