Please login to your online account to display your discounted pricing

5-Methylpyrimidine, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 5-Methylpyrimidine
Assay Percent Range 98%
Molecular Formula C5H6N2
Formula Weight 94.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC450730010

 
thermo fisher scientific
AC450730010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450732500

 
thermo fisher scientific
AC450732500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 5-Methylpyrimidine
Quantity 1g
CAS 2036-41-1
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C5H6N2
Formula Weight 94.11