Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylpyrimidine, 97+%

Chemical Identifiers

Chemical Name or Material 5-Methylpyrimidine
CAS 2036-41-1
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00047386
UN Number UN1325
Beilstein 105684
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1963803

  SDS
alfa aesar™
L1963803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19638MD

  SDS
alfa aesar™
L19638MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 94.12
Chemical Name or Material 5-Methylpyrimidine
Assay ≥97%
Molecular Formula C5H6N2
MDL Number MFCD00047386
CAS 2036-41-1
UN Number UN1325
Boiling Point 152° to 154°C
Flash Point 53°C (127°F)
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
TSCA TSCA
Beilstein 105684
Melting Point 29° to 31°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.