Learn More
5-Methylpyridine-2-carbonitrile, ≥97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6453914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Methylpyridine-2-carbonitrile | |
72°C to 75°C | |
MFCD06200830 | |
UN3439 | |
2-cyano-5-methylpyridine, 5-methylpicolinonitrile, 5-methyl-pyridine-2-carbonitrile, 5-methyl-2-pyridinecarbonitrile, 6-cyano-3-picoline, 2-pyridinecarbonitrile, 5-methyl, 2-cyano-5-methylpyridine 2-cyano-5-picoline, 5-methypicolinonitrile, pubchem12978, 5-methyl picolinonitrile | |
CC1=CC=C(N=C1)C#N | |
118.14 | |
118.14 | |
Solid |
1620-77-5 | |
C7H6N2 | |
25 g | |
111125 | |
LIEQVZZZYLHNRH-UHFFFAOYSA-N | |
5-methylpyridine-2-carbonitrile | |
74183 | |
≥97% |
Chemical Identifiers
1620-77-5 | |
118.14 | |
LIEQVZZZYLHNRH-UHFFFAOYSA-N | |
74183 | |
CC1=CC=C(N=C1)C#N |
C7H6N2 | |
MFCD06200830 | |
2-cyano-5-methylpyridine, 5-methylpicolinonitrile, 5-methyl-pyridine-2-carbonitrile, 5-methyl-2-pyridinecarbonitrile, 6-cyano-3-picoline, 2-pyridinecarbonitrile, 5-methyl, 2-cyano-5-methylpyridine 2-cyano-5-picoline, 5-methypicolinonitrile, pubchem12978, 5-methyl picolinonitrile | |
5-methylpyridine-2-carbonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 216-589-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only