missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester, 50-70%, see CofA for e, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439950010
Specifications
5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester | |
1g | |
C16H19BN2O2 | |
AVMWJACSYAQNDG-UHFFFAOYSA-N | |
2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane | |
16217859 | |
50-70%,see CofA for e |
>300.0°C | |
872054-54-1 | |
2-5-methyl-2-pyridinyl-6-phenyl-1,3,6,2-dioxazaborocane, 2-5-methyl-2-pyridinyl-6-phenyl-5,6,7,8-tetrahydro-4h-1,3,6,2-dioxazaborocine, 2-5-methyl-pyridin-2-yl-6-phenyl-1,3,6,2 dioxazaborolane, 2-5-methylpyridin-2-yl-6-phenyl-1,3,6,2-dioxazaborocane, 5-methyl-2-pyridineboronic acid n-phenyldiethanolamine ester, 5-methylpyridine-2-boronic acid n-phenyldiethanolamine ester | |
B1(OCCN(CCO1)C2=CC=CC=C2)C3=NC=C(C=C3)C | |
282.15 | |
282.15 |
Chemical Identifiers
872054-54-1 | |
282.15 | |
2-5-methyl-2-pyridinyl-6-phenyl-1,3,6,2-dioxazaborocane, 2-5-methyl-2-pyridinyl-6-phenyl-5,6,7,8-tetrahydro-4h-1,3,6,2-dioxazaborocine, 2-5-methyl-pyridin-2-yl-6-phenyl-1,3,6,2 dioxazaborolane, 2-5-methylpyridin-2-yl-6-phenyl-1,3,6,2-dioxazaborocane, 5-methyl-2-pyridineboronic acid n-phenyldiethanolamine ester, 5-methylpyridine-2-boronic acid n-phenyldiethanolamine ester | |
2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane |
C16H19BN2O2 | |
AVMWJACSYAQNDG-UHFFFAOYSA-N | |
16217859 | |
B1(OCCN(CCO1)C2=CC=CC=C2)C3=NC=C(C=C3)C |
Safety and Handling
GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a P
GHS Signal Word: Warning