missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester, 50-70%, see CofA for e, ACROS Organics™

Chemical Identifiers

CAS 872054-54-1
Molecular Formula C16H19BN2O2
Molecular Weight (g/mol) 282.15
InChI Key AVMWJACSYAQNDG-UHFFFAOYSA-N
Synonym 2-5-methyl-2-pyridinyl-6-phenyl-1,3,6,2-dioxazaborocane, 2-5-methyl-2-pyridinyl-6-phenyl-5,6,7,8-tetrahydro-4h-1,3,6,2-dioxazaborocine, 2-5-methyl-pyridin-2-yl-6-phenyl-1,3,6,2 dioxazaborolane, 2-5-methylpyridin-2-yl-6-phenyl-1,3,6,2-dioxazaborocane, 5-methyl-2-pyridineboronic acid n-phenyldiethanolamine ester, 5-methylpyridine-2-boronic acid n-phenyldiethanolamine ester
PubChem CID 16217859
IUPAC Name 2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane
SMILES B1(OCCN(CCO1)C2=CC=CC=C2)C3=NC=C(C=C3)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC439950010
SDS View Documents
Thermo Fisher Scientific
AC439950010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439950050
SDS View Documents
Thermo Fisher Scientific
AC439950050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

872054-54-1
282.15
2-5-methyl-2-pyridinyl-6-phenyl-1,3,6,2-dioxazaborocane, 2-5-methyl-2-pyridinyl-6-phenyl-5,6,7,8-tetrahydro-4h-1,3,6,2-dioxazaborocine, 2-5-methyl-pyridin-2-yl-6-phenyl-1,3,6,2 dioxazaborolane, 2-5-methylpyridin-2-yl-6-phenyl-1,3,6,2-dioxazaborocane, 5-methyl-2-pyridineboronic acid n-phenyldiethanolamine ester, 5-methylpyridine-2-boronic acid n-phenyldiethanolamine ester
2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane
C16H19BN2O2
AVMWJACSYAQNDG-UHFFFAOYSA-N
16217859
B1(OCCN(CCO1)C2=CC=CC=C2)C3=NC=C(C=C3)C

Specifications

>300.0°C
872054-54-1
2-5-methyl-2-pyridinyl-6-phenyl-1,3,6,2-dioxazaborocane, 2-5-methyl-2-pyridinyl-6-phenyl-5,6,7,8-tetrahydro-4h-1,3,6,2-dioxazaborocine, 2-5-methyl-pyridin-2-yl-6-phenyl-1,3,6,2 dioxazaborolane, 2-5-methylpyridin-2-yl-6-phenyl-1,3,6,2-dioxazaborocane, 5-methyl-2-pyridineboronic acid n-phenyldiethanolamine ester, 5-methylpyridine-2-boronic acid n-phenyldiethanolamine ester
B1(OCCN(CCO1)C2=CC=CC=C2)C3=NC=C(C=C3)C
282.15
282.15
5-Methylpyridine-2-boronic acid N-phenyldiethanolamine ester
1g
C16H19BN2O2
AVMWJACSYAQNDG-UHFFFAOYSA-N
2-(5-methylpyridin-2-yl)-6-phenyl-1,3,6,2-dioxazaborocane
16217859
50-70%,see CofA for e
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a P

GHS Signal Word: Warning

SDS