missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylpyrazine-2-carboxylic Acid, TCI America™

$24.44 - $71.81

Chemical Identifiers

CAS 5521-55-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00068241
InChI Key RBYJWCRKFLGNDB-UHFFFAOYSA-N
Synonym 5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi
PubChem CID 122831
IUPAC Name 5-methylpyrazine-2-carboxylic acid
SMILES CC1=NC=C(N=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10035G View Documents TCI America
M10035G
5g Each for $24.44
 
M100325G View Documents TCI America
M100325G
25g Each for $71.81
 
Specifications

Chemical Identifiers

5521-55-1
138.126
RBYJWCRKFLGNDB-UHFFFAOYSA-N
122831
CC1=NC=C(N=C1)C(=O)O
C6H6N2O2
MFCD00068241
5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi
5-methylpyrazine-2-carboxylic acid

Specifications

Undesignated
5g
C6H6N2O2
5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi
CC1=NC=C(N=C1)C(=O)O
138.126
138.13
≥98.0% (GC,T)
166°C
5521-55-1
MFCD00068241
RBYJWCRKFLGNDB-UHFFFAOYSA-N
5-methylpyrazine-2-carboxylic acid
122831
Crystal-Powder at 20°C
5-Methylpyrazine-2-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No