missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Methylisoxazole-3-carboxylic Acid 98.0+%, TCI America™

Manufacturer:  Tci America M240525G

 View more versions of this product

Catalog No. M240525G



Specifications

Specifications

5-Methylisoxazole-3-carboxylic Acid
174°C
3405-77-4
MFCD01318162
BNMPIJWVMVNSRD-UHFFFAOYSA-N
5-methyl-1,2-oxazole-3-carboxylic acid
76947
Crystal-Powder at 20°C
White-Yellow
25g
C5H5NO3
3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylic acid, 5-methyl, 3-isoxazolecarboxylicacid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, isoxazole acid, ksc224c5p, pubchem15552
CC1=CC(=NO1)C(=O)O
127.099
127.10
≥98.0% (HPLC,T)

Chemical Identifiers

3405-77-4
127.099
BNMPIJWVMVNSRD-UHFFFAOYSA-N
76947
CC1=CC(=NO1)C(=O)O
C5H5NO3
MFCD01318162
3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylic acid, 5-methyl, 3-isoxazolecarboxylicacid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, isoxazole acid, ksc224c5p, pubchem15552
5-methyl-1,2-oxazole-3-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions