missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylhydantoin, 98%

$61.19 - $188.49

Chemical Identifiers

CAS 616-03-5
Molecular Formula C4H6N2O2
Formula Weight 114.1
MDL Number MFCD00022398
Beilstein 81081
Chemical Name or Material 5-Methylhydantoin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1199203 SDS Alfa Aesar™
L1199203
1g Each for $61.19
AAL1199206 SDS Alfa Aesar™
L1199206
5g Each for $188.49
Specifications

Description

Applications
5-Methylhydantoin is usede as a reactant for preparation of N-chlorohydantoin, Mitsunobu reaction, Pseudomonas putida strain allowing conversion into L-amino acids, D-hydantoinase from Agrobacteria tumefaceins and enzymatic racemization.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

616-03-5
114.1
81081
C4H6N2O2
MFCD00022398
5-Methylhydantoin

Specifications

150° to 152°C
616-03-5
114.1
81081
98%
1g
C4H6N2O2
MFCD00022398
5-Methylhydantoin