missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylhydantoin, 98%

Manufacturer:  Alfa Aesar™ L1199206

 View more versions of this product

Catalog No. AAL1199206



Description

Description

Applications
5-Methylhydantoin is usede as a reactant for preparation of N-chlorohydantoin, Mitsunobu reaction, Pseudomonas putida strain allowing conversion into L-amino acids, D-hydantoinase from Agrobacteria tumefaceins and enzymatic racemization.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

5-Methylhydantoin
5g
C4H6N2O2
81081
VMAQYKGITHDWKL-UHFFFAOYSA-N
5-methylimidazolidine-2,4-dione
69216
98%
150°C to 152°C
616-03-5
MFCD00022398
5-methylhydantoin, hydantoin, 5-methyl, 2,4-imidazolidinedione, 5-methyl, 5-methyl-2,4-imidazolidinedione, d-5-methylhydantoin, 5-methyl-hydantoin, spectrum_001906, 2, 5-methyl, spectrum3_000916, spectrum4_001200
CC1C(=O)NC(=O)N1
114.104
114.1

Chemical Identifiers

616-03-5
114.104
VMAQYKGITHDWKL-UHFFFAOYSA-N
69216
CC1C(=O)NC(=O)N1
C4H6N2O2
MFCD00022398
5-methylhydantoin, hydantoin, 5-methyl, 2,4-imidazolidinedione, 5-methyl, 5-methyl-2,4-imidazolidinedione, d-5-methylhydantoin, 5-methyl-hydantoin, spectrum_001906, 2, 5-methyl, spectrum3_000916, spectrum4_001200
5-methylimidazolidine-2,4-dione
Documents