Learn More
5-Methylhexanoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1901218
Description
Ion-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIon-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
5-Methylhexanoic acid | |
0.91 | |
102°C (216°F) | |
C7H14O2 | |
MFCD00036494 | |
UN3265 | |
Slightly soluble in water. | |
CC(C)CCCC(O)=O | |
130.19 | |
130.19 |
628-46-6 | |
216°C | |
Rancid | |
1.422 | |
50 g | |
isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2 | |
MHPUGCYGQWGLJL-UHFFFAOYSA-N | |
5-methylhexanoic acid | |
12344 | |
98% |
Chemical Identifiers
628-46-6 | |
130.19 | |
MHPUGCYGQWGLJL-UHFFFAOYSA-N | |
12344 | |
CC(C)CCCC(O)=O |
C7H14O2 | |
MFCD00036494 | |
isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2 | |
5-methylhexanoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber : MO8400900
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only