missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methylfurfurylamine 98.0+%, TCI America™
Supplier: TCI America M131125G
Specifications
5-Methylfurfurylamine | |
Yellow | |
C6H9NO | |
25 g | |
5-methylfurfurylamine, 5-methylfuran-2-yl methanamine, 5-methyl-2-furanmethanamine, 2-aminomethyl-5-methylfuran, 5-methyl-2-furyl methylamine, 2-furanmethanamine, 5-methyl, 5-methyl-2-furyl methyl amine, 1-5-methylfuran-2-yl methanamine, pubchem6995, acmc-209cjz | |
CC1=CC=C(CN)O1 | |
111.14 | |
111.14 | |
Liquid |
14003-16-8 | |
55°C | |
MFCD00143471 | |
2733 | |
YSEAGSCGERFGBL-UHFFFAOYSA-N | |
1-(5-methylfuran-2-yl)methanamine | |
2724683 | |
≥98.0% (GC,T) |
Chemical Identifiers
14003-16-8 | |
111.14 | |
YSEAGSCGERFGBL-UHFFFAOYSA-N | |
2724683 | |
CC1=CC=C(CN)O1 |
C6H9NO | |
MFCD00143471 | |
5-methylfurfurylamine, 5-methylfuran-2-yl methanamine, 5-methyl-2-furanmethanamine, 2-aminomethyl-5-methylfuran, 5-methyl-2-furyl methylamine, 2-furanmethanamine, 5-methyl, 5-methyl-2-furyl methyl amine, 1-5-methylfuran-2-yl methanamine, pubchem6995, acmc-209cjz | |
1-(5-methylfuran-2-yl)methanamine |
Safety and Handling
TSCA : No