missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methylfurfural, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 159920250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
5-Methylfurfural | |
620-02-0 | |
98.0 | |
Yellow to Brown | |
187°C | |
Authentic | |
Glass bottle | |
1.5290 to 1.532 | |
25 g | |
1.1 | |
Solubility in water: 33.3g/L (20 c) | |
CC1=CC=C(O1)C=O | |
110.11 | |
CHEBI:2091 | |
98+% |
98+% | |
10mg KOH/g max. | |
100.0 | |
1.1000g/mL | |
72°C | |
98% min. (GC) | |
C6H6O2 | |
MFCD00003232 | |
17, 289 | |
5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
OUDFNZMQXZILJD-UHFFFAOYSA-N | |
5-methylfuran-2-carbaldehyde | |
12097 | |
110.11 | |
Liquid |
Chemical Identifiers
620-02-0 | |
110.11 | |
OUDFNZMQXZILJD-UHFFFAOYSA-N | |
12097 | |
5-methylfuran-2-carbaldehyde |
C6H6O2 | |
MFCD00003232 | |
5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran | |
CHEBI:2091 | |
CC1=CC=C(O1)C=O |
Safety and Handling
EINECSNumber : 210-622-6
RTECSNumber : LT7032500
TSCA : TSCA
RUO – Research Use Only