missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 5-Methylcytosine, 97%

Manufacturer:  Alfa Aesar™ 43492MC

 View more versions of this product

Catalog No. AA43492MC



Specifications

Specifications

5-Methylcytosine
554-01-8
MFCD00233537
2 1h-pyrimidinone, 4-amino-5-methyl
CC1=C(NC(=O)N=C1)N
125.131
CHEBI:27551
Crystalline
100mg
C5H7N3O
14,6050
LRSASMSXMSNRBT-UHFFFAOYSA-N
6-amino-5-methyl-1H-pyrimidin-2-one
65040
125.13
97%

Chemical Identifiers

554-01-8
125.131
LRSASMSXMSNRBT-UHFFFAOYSA-N
65040
6-amino-5-methyl-1H-pyrimidin-2-one
C5H7N3O
MFCD00233537
2 1h-pyrimidinone, 4-amino-5-methyl
CHEBI:27551
CC1=C(NC(=O)N=C1)N
Safety and Handling

Safety and Handling

RTECSNumber : UW7362350

Documents